Ethylenediamine, 99%
Ethylenediamine, 99%
Ethylenediamine, 99%
Have Questions?
Catalog NumberQuantity
A12132.0D10,000 mL
Catalog number A12132.0D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10,000 mL
Request bulk or custom format
Specifications
Melting Point-11°C
Boiling Point117°C to 118°C
Quantity10,000 mL
Percent Purity99%
Physical FormLiquid
View more
Ethylenediamine is a strongly basic amine useful as a building block in chemical synthesis. It is used as a solvent to dissolve proteins such as albumins and casein. It is widely used for color photography developers, binders, adhesives, fabric softeners, curing agents for epoxys and dyes. As a corrosion inhibitor, it plays a vital role in paints and coolants. It is used as an intermediate in the preparation of polyamide resins, fuel additives and lubricants. It acts as a precursor for many polymers like polyurethane fibers and poly(amidoamine), ethylenediamine dihydroiodide (EDDI) as well as the bleaching activator, tetraacetylethylenediamine. It is an important chelating ligand used in the preparation of coordination compounds viz. tris(ethylenediamine)cobalt(III) chloride. It is also involved in the manufacture of many industrial chemicals and forms derivatives with carboxylic acids, nitriles, alcohols, alkylating agents, carbon disulfide, aldehydes and ketones. It is a basic building block to prepare heterocyclic compound such as imidazolidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Strongly basic amine useful as a building block in chemical synthesis