Owl™ A1 Large Gel System
Owl™ A1 Large Gel System
Actual product may vary
Owl™ A1 Large Gel System
Owl™ A1 Large Gel System
Owl™ A1 Large Gel System
Owl™ A1 Large Gel System
Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems, which have smaller footprints ideal for detailed RNA/DNA analysis.
Have Questions?
Catalog NumberType
A1-BPLarge, w/Buffer Exchange Ports
Catalog number A1-BP
Price (SGD)
-
Type:
Large, w/Buffer Exchange Ports
Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems. With one of the smallest footprints of any large-format device, these systems are ideal for detailed RNA/DNA analysis.
Features:
  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • Programmable power inverter may be used for field reversal electrophoresis
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Includes:
  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (12-, 16- and 20-well), 1.5mm thick
Specifications
Certifications/ComplianceCE marked
DescriptionComplete system with buffer exchange ports
Product LineOwl
TypeLarge, w/Buffer Exchange Ports
Sample Size8 to 150
Volume (Metric) Buffer1.6 L
Dimensions (L x W x H)14.8 x 8.5 x 4.3 in. (37.5 x 21.5 x 11 cm)
Length (English)14.8 in.
Length (Metric)37.5 cm
Length (Metric) Gel25 cm
Width (English)8.5 in.
Width (Metric)21.5 cm
Width (Metric) Gel13 cm
Height (English)4.3 in.
Height (Metric)11 cm
Unit SizeEA