Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Actual product may vary
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Thermo Scientific
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Provide uncompromised sample protection for -50° to -86°C applications with Thermo ScientificTM FormaTM 900 Series -86°C Upright Ultra-Low Temperature Freezers.
Have Questions?
Catalog NumberElectrical RequirementsHoldsCapacity (English)No. of Doors
995
also known as 995SS
230 V 50 HzBoxes (360)23 cu. ft.2
Catalog number 995
also known as 995SS
HSN 84184090
Price (SGD)
-
Electrical Requirements:
230 V 50 Hz
Holds:
Boxes (360)
Capacity (English):
23 cu. ft.
No. of Doors:
2
Provide uncompromised sample protection for -50° to -86°C applications with Thermo ScientificTM FormaTM 900 Series -86°C Upright Ultra-Low Temperature Freezers. Available in single or double door models, these freezers are designed for daily sample protection and dependability.
Key Features
  • Centralized, eye level information center, including microprocessor control and monitoring system; all controls and displays are easy to reach and read
  • Improves temperature control and increases reserve BTUH capacity, resulting in a more stable temperature sample
  • Unique double door to store frequently used samples in the top chamber while long-term storage remains undisturbed in bottom
  • Single-hand operation: easy-to-use, padlock-compatible, ergonomic door handle with integrated key lock
  • Optional CO2 or LN2 back-up: Choice of CO2 or LN2 safety back-up systems for additional sample protection in the event of a power or mechanical failure
  • Optional chart recorder: Built-in chart recorder can be installed at eye level on Forma 900 -86°C models
Construction:
  • 5 in. (127mm) foamed-in-place, polyurethane insulation: Reduces power consumption, maintains temperature setpoint
  • Triple-point door gasket, provides longer holding time in the event of a door opening
  • Rugged, heavy gauge, cold-rolled steel cabinets resist chipping and rust
  • 1 in. (25mm) access ports: Allow for the use of inexpedient probes or instrumentation
  • Four inner doors reduce cold air loss and improve temperature recovery after door closings
  • Painted Galvaneel interior (stainless steel option available)
  • Water-cooled condenser option available
Warranty varies by country, please contact us for details
Specifications
DescriptionForma Series Ultra-Low Freezer, 360-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)23 cu. ft.
Capacity (Metric)651 L
Temperature Range-50°C to -86°C
Door StyleOuter (1), Inner (4)
No. of Doors2
Inner Doors4
Cabinet MaterialCold-rolled Steel
InsulationPolyurethane Foam
InteriorPainted Galvaneel - Standard, Stainless-steel - Optional
Shelves3
Capacity (English) Shelf150 lb.
Capacity (Metric) Shelf68 kg
Certifications/ComplianceCE, cULus, FDA Class I, UL
StyleUpright
DisplayPush Button LED
Condenser TypeFin and Tube, Forced-Air Cooled
Product LineForma
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Water CooledOptional
Dimensions (DxWxH) Exterior38.9 x 40.8 x 77.9 in. (98.9 x 103.6 x 197.9 cm)
Dimensions (D x W x H) Interior25.3 x 30.6 x 51.5 in. (64.3 x 77.7 x 130.8 cm)
Footprint (English)11.02 sq. ft.
Footprint (Metric)1.02 sq. meters
Monitoring OptionsAlarm, Recorder
Access SecurityKeyLock, Padlock Standard
Chart Recorder7-day Ink or Inkless (Optional)
Data OutputsRS-485, 4-20mA, dry contacts - standard, PT100 probe Optional
On-Board DataloggingNot Available
Operating ModesStandard Operation
Setpoint SecurityNot Available
Backup SystemsCO2 or LN2 Optional
Smart View CompatibilitySmart-Vue Compatible
Amperage12 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Wattage980 W
Energy Usage19kW/day
Line Voltage IndicatorBuck/Boost
Voltage CompensationBuck/Boost
Vial Capacity36,000 x 2mL
CompressorHermetic
RefrigerantR404A (1st stage), R508B (2nd stage)
Shipping Weight (English)900 lb.
Shipping Weight (Metric)408 kg
Weight (English)850 lb.
Weight (Metric)385 kg
HoldsBoxes (360)
Unit SizeEach
Frequently asked questions (FAQs)
The low battery alarm is displaying on my ULT (Ultra-Low Temperature) freezer. Can I buy a new battery?
You can order a new battery (Cat. No. 400159) online.