Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Actual product may vary
Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.
Have Questions?
Catalog NumberVolume (Metric)FilteredSterilityFit for Purpose
966-261300 μLNon-filteredSterileSterile
Catalog number 966-261
Price (SGD)
996.00
CS
Add to cart
Volume (Metric):
300 μL
Filtered:
Non-filtered
Sterility:
Sterile
Fit for Purpose:
Sterile
Price (SGD)
996.00
CS
Add to cart
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
Volume (Metric)300 μL
FilteredNon-filtered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizeCS