Check Order Status
Reagent Reservoir
Actual product may vary
Reagent Reservoir
The reagent reservoir from Thermo Electron LED GmbH is made of polypropylene and has eight wells.
Have Questions?
Catalog number
9510037
Price (SGD)
51.00
PK
Add to cart
Price (SGD)
51.00
PK
Add to cart
The reagent reservoir from Thermo Electron LED GmbH is made of polypropylene and has eight wells.
Autoclavable
Provide variety of solutions to suit all pipetting procedures
Reagent reservoirs designed to provide solutions tailored to procedures
Specifications
Color
White
Capacity (Metric)
60 mL
Material
Polypropylene
Sterility
Non-sterile
Type
Reagent Reservoir
Quantity
10 Pk.
Autoclavable
Autoclavable
Channels
8
Disposable
Reusable
Bottom Shape
Flat