Reagent Reservoirs
Reagent Reservoirs
Actual product may vary
Reagent Reservoirs
Reagent Reservoirs
Reagent Reservoirs
Reagent Reservoirs
Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MaterialSterilityPackaging
951002760 mLPolypropyleneNon-sterileBulk
9512809525 mLPolystyreneNon-sterile10 Bags of 10
8096-1125 mLPlasticSterileIndividually Wrapped
8093-1125 mLPolystyreneSterile10 Bags of 10
95128085100 mLPolystyreneNon-sterile10 Bags of 10
9512809325 mLPolystyreneNon-sterileBulk
Catalog number 9510027
Price (SGD)
28.00
PK
Add to cart
Capacity (Metric):
60 mL
Material:
Polypropylene
Sterility:
Non-sterile
Packaging:
Bulk
Price (SGD)
28.00
PK
Add to cart
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Benefits
  • Trough within a trough maximizes the amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25mL Divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
Specifications
Capacity (English)2 oz.
ColorWhite
Capacity (Metric)60 mL
MaterialPolypropylene
SterilityNon-sterile
TypeReagent Reservoir
PackagingBulk