ClipTip™ Pipette Tips
ClipTip™ Pipette Tips
Actual product may vary
ClipTip™ Pipette Tips
ClipTip™ Pipette Tips
ClipTip™ Pipette Tips
Thermo Fisher
ClipTip™ Pipette Tips
Ensure a secure seal for every sample, with these pipette tips. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFormatTip Style
9441025050 μLNon-sterileRackLow Retention
9441004312.5 μLSterileRackStandard
944108101250 μLNon-sterileRackLow Retention
944108131250 μLSterileRackLow Retention
94410210
also known as FS94410210
20 μLNon-sterileRackLow Retention
94410310200 μLNon-sterileRackLow Retention
94410510
also known as FS94410510
300 μLNon-sterileRackLow Retention
FIS14-488-044300 μLNon-sterileRackLow Retention
94410710
also known as FS94410710
1000 μLNon-sterileRackLow Retention
9441021820 μLSterileReload DeckLow Retention
9441025850 μLSterileReload DeckLow Retention
94410318200 μLSterileReload DeckLow Retention
94410518300 μLSterileReload DeckLow Retention
944107181000 μLSterileReload DeckLow Retention
94410217
also known as FS94410217
20 μLNon-sterileReload DeckLow Retention
94410317
also known as FS94410317
200 μLNon-sterileReload DeckLow Retention
94410517
also known as FS94410517
300 μLNon-sterileReload DeckLow Retention
9441021320 μLSterileRackLow Retention
94410253
also known as FS94410253
50 μLSterileRackLow Retention
94410313200 μLSterileRackLow Retention
FIS14-488-058200 μLSterileRackLow Retention
94410513
also known as FS94410513
300 μLSterileRackLow Retention
Catalog number 94410250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
50 μL
Sterility:
Non-sterile
Format:
Rack
Tip Style:
Low Retention
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.
Key Features:
  • ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
  • Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
  • ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
Low retention
  • Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
  • ClipTip 20-1250 96-format tips, inserts and racks can be autoclaved at 121°C (252°F) (2 ata) for 20 minutes. Tips in reload packages can be autoclaved when loaded to an empty rack. Filtered tips are not autoclavable.
Environmentally responsible
  • ClipTip reload system uses recycled components and 30% less plastic
Versatile Tip Range
  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended lenght tips available for volumes 12.5 and 300 μl allowing for better reach with specific labware consumables
Immediate traceability
  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionClipTip 50, Non-filtered, Non-sterile, Rack
Volume (Metric)50 μL
FilteredNon-filtered
SterilityNon-sterile
AutoclavableAutoclavable
Length (English)2.01 in.
Length (Metric)5.1cm
FormatRack
Tip StyleLow Retention
Packaging10 x 96 tips/rack
Color CodeViolet
For Use With (Equipment)Thermo Scientific F1-ClipTip
Product LineClipTip
Fit for PurposePCR Ready
Unit SizePK