Finntip™ Flex™ Pipette Tips
Actual product may vary
Thermo Scientific
Finntip™ Flex™ Pipette Tips
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
Have Questions?
Catalog NumberVolume (Metric)Color CodePackagingFit for Purpose
940608131200 μLTurquoise960 tipsSterile & PCR Ready
Catalog number 94060813
HSN 84799090
Price (SGD)
302.00
Pack
Volume (Metric):
1200 μL
Color Code:
Turquoise
Packaging:
960 tips
Fit for Purpose:
Sterile & PCR Ready
Price (SGD)
302.00
Pack
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Key Features:
  • Excellent for single and multichannel pipetting
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized, 10 x 96 rack
Softer Finntip Flex Tip
  • The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Available in six volume ranges: 0.2-10 μL, 1-200 μL, 5-300 μL, 100-1000 μL, 50-1200 μL and 10mL Extended (bottle tip)
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip Flex 1200, Non-Filtered, Sterile, Rack
Volume (Metric)1200 μL
FilteredNon-filtered
SterilitySterile
ColorTurquoise
ModelFinntip Flex 1200
Length (English)3.4 in.
Length (Metric)8.7cm
FormatRack
Packaging960 tips
Color CodeTurquoise
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip Flex
Fit for PurposeSterile & PCR Ready
Unit SizePack