Finntip™ Biocon Pipette Tips
Finntip™ Biocon Pipette Tips
Actual product may vary
Finntip™ Biocon Pipette Tips
Perform PCR and other purity demanding applications with these biocon pipette tips, certified free from endotoxin, DNA, DNase and RNase.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
940533001000 μL
9405310010 μL
94053200250 μL
Catalog number 94053300
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1000 μL
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Features:
  • Individually packed
  • Three models in volume range 0.2-1000 μL
  • Presterilized with gamma radiation
  • Includes certificate of quality
Recommended for: PCR applications, molecular biology
Specifications
DescriptionFinntip BioCon 1000, Non-Filtered, Sterile,
Volume (Metric)1000 μL
FilteredNon-filtered
SterilitySterile
ModelFinntip BioCon 1000
Certifications/ComplianceISO 9001, ISO 14001, ISO 13485
For Use With (Equipment)Thermo Scientific FinnTip BioCon 1000
Product LineFinnTip Biocon
Unit SizePK