Finntip™ Filtered Pipette Tips
Actual product may vary
Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Protect pipettes and the samples from contamination with these filtered pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)
940525505 mL14.7 cm
9405200010 μL3.2 cm
FIS21-377-35410 μL3.2 cm
9405202020 μL3.3 cm
9405206050 μL4.8 cm
FIS21-377-38750 μL4.8 cm
9405210010 μL5.2 cm
FIS21-377-16910 μL5.2 cm
9405215020 μL5.2 cm
FIS21-377-35320 μL5.2 cm
9405216030 μL5.2 cm
94052200100 μL5.2 cm
FIS21-377-170100 μL5.2 cm
94052300200 μL5.2 cm
FIS21-377-171200 μL5.2 cm
94052310100 μL7.8 cm
FIS14-386-54100 μL7.8 cm
FKS11428529100 μL7.8 cm
94052320200 μL7.8 cm
94052350300 μL5.2 cm
940524101000 μL7.1 cm
FIS21-377-6041000 μL7.1 cm
FKS117498551000 μL7.1 cm
940524301000 μL10.5 cm
FIS21-377-6051000 μL10.5 cm
9405280010 mL15 cm
94052450150 to 1500 μL6.99 cm
FIS21-377-5125 mL14.7 cm
Catalog number 94052550
HSN 84799090
Price (SGD)
235.00
Special offer
Ends: 30-Jun-2026
261.00
Save 26.00 (10%)
Pack
Volume (Metric):
5 mL
Length (Metric):
14.7 cm
Price (SGD)
235.00
Special offer
Ends: 30-Jun-2026
261.00
Save 26.00 (10%)
Pack
Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0.2μL-10mL.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Filtered Finntip pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
  • Inert and non-sealing polyethylene filter
  • Tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 20, 30, 100, 200, 300 and 1000 μL
  • Extended length tips available for volumes 100, 200 and 1000 μL allowing for better reach with specific labware consumables
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionFinntip Filter 5 mL, sterile
Volume (Metric)5 mL
FilteredFiltered
SterilitySterile
AutoclavableNot Autoclavable
Length (Metric)14.7 cm
FormatRack
Color CodeGreen
DisposableSingle-use
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip
Fit for PurposeSterile & PCR Ready
Unit SizePack