Finntip™ Pipette Tips
Actual product may vary
Finntip™ Pipette Tips
Finntip™ Pipette Tips
Finntip™ Pipette Tips
Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFit for PurposeColor Code
94011151000 μLNon-sterileCertified QualityBlue
940030010 μLNon-sterileCertified QualityPink
FIS21-377-20810 μLNon-sterileCertified QualityPink
940030310 μLSterileSterile & PCR ReadyPink
940031010 μLNon-sterileCertified QualityPink
FIS21-377-20910 μLNon-sterileCertified QualityPink
940032710 μLNon-sterileCertified QualityPink
94010301000 μLNon-sterileCertified QualityBlue
FIS21-377-2991000 μLNon-sterileCertified QualityBlue
94011101000 μLNon-sterileCertified QualityBlue
94011131000 μLSterileSterile & PCR ReadyBlue
9401250300 μLNon-sterileCertified QualityOrange
9401253300 μLSterileSterile & PCR ReadyOrange
9401255300 μLNon-sterileCertified QualityOrange
9401260300 μLNon-sterileCertified QualityOrange
94020305 mLNon-sterileCertified QualityGreen
FKS102176205 mLNon-sterileCertified QualityGreen
94020705 mLNon-sterileCertified QualityGreen
FIS21-377-5105 mLNon-sterileCertified QualityGreen
94020735 mLSterileSterile & PCR ReadyGreen
940217110 mLNon-sterileCertified QualityRed
FKS1010538010 mLNon-sterileCertified QualityRed
940218010 mLNon-sterileCertified QualityRed
940218310 mLSterileSterile & PCR ReadyRed
940036050 μLNon-sterileCertified QualityTurquoise
940037050 μLNon-sterileCertified QualityTurquoise
FIS21-377-38550 μLNon-sterileCertified QualityTurquoise
940037350 μLSterileSterile & PCR ReadyTurquoise
Catalog number 9401115
HSN 84799090
Price (SGD)
108.00
Special offer
Ends: 30-Jun-2026
120.00
Save 12.00 (10%)
Pack
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Fit for Purpose:
Certified Quality
Color Code:
Blue
Price (SGD)
108.00
Special offer
Ends: 30-Jun-2026
120.00
Save 12.00 (10%)
Pack
Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 250, 300 and 1000 μL
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 1000 Refill pack, Non-Filtered, Non-sterile, Refill
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlue
ModelFinntip 1000 Refill pack
Length (English)2.8 in.
Length (Metric)7.1cm
FormatRefill
Color CodeBlue
For Use With (Equipment)Thermo Scientific Finnpipette 1000
Product LineFinnTip
Fit for PurposeCertified Quality
Unit SizePack