Finntip™ Non-Filtered 384 Format Pipette Tips 20 μL
Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Actual product may vary
Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Thermo Scientific
Finntip™ Non-Filtered 384 Format Pipette Tips 20 μL
Ensure accurate, precise pipetting. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFit for PurposeFormat
9400613SterileSterile & PCR ReadyRack
9400620Non-sterileCertified QualityBulk
9400610Non-sterileCertified QualityRack
Catalog number 9400613
HSN 84799090
Price (SGD)
1,028.00
PK
Add to cart
Sterility:
Sterile
Fit for Purpose:
Sterile & PCR Ready
Format:
Rack
Price (SGD)
1,028.00
PK
Add to cart
Accurate, precise pipetting with 384-format volume 20 μL. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 20 Sterile, Non-Filtered, Sterile, Rack
Volume (Metric)20 μL
FilteredNon-filtered
SterilitySterile
ColorPurple
AutoclavableFinntip pipette tips and racks can be autoclaved for 121 degrees Celsius for 20 minutes
ModelFinntip 20 Sterile
Length (English)1.3 in.
Length (Metric)3.3cm
FormatRack
Color CodePurple
For Use With (Equipment)Thermo Scientific Finnpipette 20
Product LineFinnTip
Fit for PurposeSterile & PCR Ready
Shelf LifeRecommended shelf life for Finntip pipette tips is five years
Unit SizePK