Finntip™ Non-Filtered Universal Pipette Tips
Finntip™ Non-Filtered Universal Pipette Tips
Actual product may vary
Finntip™ Non-Filtered Universal Pipette Tips
Finntip™ Non-Filtered Universal Pipette Tips
Finntip™ Non-Filtered Universal Pipette Tips
Ensure accurate, precise pipetting with these non-filtered, universal pipette tips, designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFormatFit for Purpose
9400267Non-sterileRefillCertified Quality
9400263SterileRackSterile and PCR Ready
FIS21-377-126SterileRackSterile and PCR Ready
Catalog number 9400267
Price (SGD)
213.00
PK
Add to cart
Sterility:
Non-sterile
Format:
Refill
Fit for Purpose:
Certified Quality
Price (SGD)
213.00
PK
Add to cart
Ensure accurate, precise pipetting with Thermo Scientific™ Finntip™ Non-Filtered Universal Pipette Tips, manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips to prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 250, 300 and 1000 μL
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 250 Universal refill kit, Non-Filtered, Non-sterile, Refill
Volume (Metric)250 μL
FilteredNon-filtered
SterilityNon-sterile
ColorYellow
ModelFinntip 250 Universal Refill Kit
Length (English)2 in.
Length (Metric)5.2 cm
FormatRefill
Tip StyleUniversal Fit
Quantity20 x 96 tips
Color CodeYellow
For Use With (Equipment)Thermo Scientific™ Finnpipette™ 250
Product LineFinnTip
Fit for PurposeCertified Quality
Unit SizePK