Glass Rolled Rim Vials
Glass Rolled Rim Vials
Actual product may vary
Glass Rolled Rim Vials
Glass Rolled Rim Vials are ideal for a variety of laboratory applications. The vials are made of soda-lime glass with a tight closing polyethylene snap-on lid, non-sterile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) Outer
940026030 mL76 mm28 mm
940023025 mL51 mm30 mm
Catalog number 9400260
Price (SGD)
125.00
PK
Add to cart
Volume (Metric):
30 mL
Height (Metric):
76 mm
Diameter (Metric) Outer:
28 mm
Price (SGD)
125.00
PK
Add to cart
  • Soda-lime glass
  • PE snap-on lid
  • Non-sterile
Specifications
Closure ColorNatural
DescriptionRolled rim glass vials with snap-on lid
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialSoda Lime Glass
SterilityNon-sterile
Volume (Metric)30 mL
For Use With (Application)A variety of laboratory applications
Height (English)3 in.
Material (Seal)Polyethylene
Neck StyleRolled Rim
Volume (English)1 oz.
Closure MaterialPolyethylene
Closure TypeSnap Cap
Height (Metric)76 mm
TypeRolled Rim Vial
Unit SizePK