Specialty Racks
Specialty Racks
Actual product may vary
Specialty Racks
Specialty Racks
Specialty Racks
Thermo Scientific
Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsPositionsProduct TypeFor Use With (Equipment)
920096Boxes (2 in.)Box Racks - Sliding Drawer17.3, 23 cu. ft. Upright Freezers
920017Boxes (3 in.)12 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920018Boxes (2 in.)20 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920022Boxes (2 in.)22 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920094Boxes (2 in.)Box Racks - Sliding Drawer13 cu. ft. Upright Freezers
920097Boxes (3 in.)12 Boxes/RackBox Racks - Sliding Drawer17.3, 23 cu. ft. Upright Freezers
398329Boxes (2 in.)20 Boxes/RackBox Racks - Sliding DrawerThermo Scientific TDE and TSX Series (5 shelves)
Catalog number 920096
HSN 84189900
Price (SGD)
196.00
EA
Add to cart
Holds:
Boxes (2 in.)
Product Type:
Box Racks - Sliding Drawer
For Use With (Equipment):
17.3, 23 cu. ft. Upright Freezers
Price (SGD)
196.00
EA
Add to cart
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
  • Warranty varies by country, please contact us for details
Specifications
Product TypeBox Racks - Sliding Drawer
ColorSilver
HoldsBoxes (2 in.)
ShapeSquare
Shelves5
For Use With (Equipment)17.3, 23 cu. ft. Upright Freezers
Width (Metric)14 cm
Width (English)5.5 in.
Height (Metric)55.9 cm
Height (English)22 in.
Depth (English)11.9 in.
Depth (Metric)30.2 cm
Unit SizeEA