Specialty Racks
Actual product may vary
Specialty Racks
Specialty Racks
Specialty Racks
Thermo Scientific
Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsPositionsProduct TypeFor Use With (Equipment)
920022Boxes (2 in.)22 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920017Boxes (3 in.)12 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920018Boxes (2 in.)20 Boxes/RackSide Access Freezer Rack (Adjustable)17.3, 23 cu. ft. Upright Freezers
920094Boxes (2 in.)Box Racks - Sliding Drawer13 cu. ft. Upright Freezers
920096Boxes (2 in.)Box Racks - Sliding Drawer17.3, 23 cu. ft. Upright Freezers
920097Boxes (3 in.)12 Boxes/RackBox Racks - Sliding Drawer17.3, 23 cu. ft. Upright Freezers
398329Boxes (2 in.)20 Boxes/RackBox Racks - Sliding DrawerThermo Scientific TDE and TSX Series
Catalog number 920022
HSN 84189900
Price (SGD)
271.00
Special offer
Ends: 30-Jun-2026
361.00
Save 90.00 (25%)
Each
Holds:
Boxes (2 in.)
Positions:
22 Boxes/Rack
Product Type:
Side Access Freezer Rack (Adjustable)
For Use With (Equipment):
17.3, 23 cu. ft. Upright Freezers
Price (SGD)
271.00
Special offer
Ends: 30-Jun-2026
361.00
Save 90.00 (25%)
Each
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
  • Warranty varies by country, please contact us for details
Specifications
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions22 Boxes/Rack
For Use With (Equipment)17.3, 23 cu. ft. Upright Freezers
Width (English)5.3 in.
Height (English)11.6 in.
Depth (English)24.6 in.
Depth (Metric)62.5 cm
Product TypeSide Access Freezer Rack (Adjustable)
Shelves5
Height (Metric)29.5 cm
Width (Metric)13.5 cm
Unit SizeEach