Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Actual product may vary
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredTip StyleFit for Purpose
919-021
also known as 919-021G
130 μLFilteredStandardSterile and PCR Ready
916-02140 μLFilteredStandardSterile and PCR Ready
Catalog number 919-021
also known as 919-021G
Price (SGD)
277.00
PK
Add to cart
Volume (Metric):
130 μL
Filtered:
Filtered
Tip Style:
Standard
Fit for Purpose:
Sterile and PCR Ready
Price (SGD)
277.00
PK
Add to cart
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Specifications
Volume (Metric)130 μL
FilteredFiltered
SterilitySterile
ColorClear
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
FormatRack
Tip StyleStandard
Packaging96 tips/rack, 10 racks/pack, 5 packs/case
Quantity4800 tips
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
Product LineBeckman, Beckman
Fit for PurposeSterile and PCR Ready
Unit SizePK