Orion™ pH Buffer Bottles
Orion™ pH Buffer Bottles
Actual product may vary
Orion™ pH Buffer Bottles
Orion™ pH Buffer Bottles
Orion™ pH Buffer Bottles
Orion™ pH Buffer Bottles
Orion™ pH Buffer Bottles
Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitypH
910110475 mL10.01 ± 0.02 pH
FIS13-643-002475 mL1.68 ± 0.01 pH
FIS13-643-0075 x 60 mL1.68 ± 0.01 pH
FIS13-079-319475 mL10.01 ± 0.02 pH
FIS09-357-685 x 60 mL10.01 ± 0.02 pH
FIS13-643-001475 mL12.46 ± 0.03 pH
FKS10114234475 mL12.46 ± 0.03 pH
FIS13-643-0055 x 60 mL12.46 ± 0.03 pH
910104475 mL4.01 ± 0.01 pH
FIS13-301-133475 mL4.01 ± 0.01 pH
FKS10759161475 mL4.01 ± 0.01 pH
FIS09-357-695 x 60 mL4.01 ± 0.01 pH
FIS13-640-8475 mL5.00 ± 0.02 pH
FIS13-643-003475 mL6.86 ± 0.01 pH
910107475 mL7.00 ± 0.01 pH
FIS13-641-857475 mL7.00 ± 0.01 pH
FKS10653951475 mL7.00 ± 0.01 pH
FIS09-357-705 x 60 mL7.00 ± 0.01 pH
FIS13-643-004475 mL9.18 ± 0.01 pH
Catalog number 910110
Price (SGD)
-
Quantity:
475 mL
pH:
10.01 ± 0.02 pH
Use Thermo Scientific™ Orion™ pH buffer bottles to perform accurate, reproducible pH calibrations on your pH meter and pH electrode measurement system. Orion pH buffers are available in 475mL bottles for everyday use and laboratory and bench pH analysis and available in 60 mL bottle 5 packs for occasional use and portable and field pH analysis. Using high-quality pH buffers to calibrate your pH system is the best way to ensure your pH measurements are correct and repeatable.
Whether you are in the lab or in the field, you can be confident that Orion solutions will deliver accurate and repeatable measurements. Orion solutions are produced in controlled batches using ultrapure water and undergo rigorous quality testing throughout the process to prevent contamination. Our strict quality procedures verify correct bottle labeling and volume, so each batch meets specific lot release criteria. Bottles are sealed and torque-tested to ensure bottle caps are secure and solutions remain free of contamination. Rigorous lifetime testing ensures sealed Orion solutions remain within published specifications for their entire shelf life and do not degrade over time.
  • Orion pH buffers in 475mL bottles are ideal for everyday use and laboratory and bench pH analysis.
  • Orion pH buffers in 60 mL bottle 5 packs are designed for occasional use and portable and field pH analysis.
  • Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards. 
  • Orion pH buffers are color-coded for easy identification: pH 4 buffer is pink, pH 5 buffer is orange, pH 7 buffer is yellow and pH 10 buffer is blue. 
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Chemical Name or MaterialpH Buffer, bottles
ColorBlue
DescriptionOrion pH 10.01 buffer, color-coded blue, 475mL bottle
For Use With (Application)Accurate pH calibrations and NIST traceability, laboratory and bench analysis
For Use With (Equipment)pH Meter and Electrode
pH10.01 ± 0.02 pH
Physical FormLiquid
Quantity475 mL
Solution TypepH Buffer
FormulationWater, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate
PackagingBottle
Traceability to NISTYes
Unit SizeEach