Product Image
Orion™ pH Electrode Storage Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Quantity
910001pH Electrode475 mL
FIS13-303-160ROSS pH Electrode475 mL
FIS13-641-908pH Electrode475 mL
FIS09-357-67pH Electrode60 mL
Catalog number 910001
HSN 38220019
Price (SGD)
-
For Use With (Equipment):
pH Electrode
Quantity:
475 mL
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.
Storage Solutions:
  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionpH electrode storage solution, 475 mL bottle
For Use With (Equipment)pH Electrode
TypepH Electrode Storage Solution
Filling Solution TypeStorage
For Use With (Application)Ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
PackagingBottle
Quantity475 mL
Unit SizeEach