Polar Silica-Based SPE Columns, Cyanopropyl (CN), 500 mg/3 mL
Product Image
Polar Silica-Based SPE Columns, Cyanopropyl (CN), 500 mg/3 mL
Medical-grade polypropylene with PE frits
Have Questions?
Catalog number 9064
Price (SGD)
133.00
EA
Add to cart
Price (SGD)
133.00
EA
Add to cart
  • All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
Specifications
Quantity50
Unit SizeEA