Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Actual product may vary
Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredFor Use With (Equipment)Fit for Purpose
904-253
also known as 904-253GPA
1000 μLNon-filteredTecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusCertified Quality
902-25150 μLNon-filteredCavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusSterile
902-25350 μLNon-filteredTecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusCertified Quality
904-011
also known as 904-011P
1000 μLFilteredTecan Freedom EVO, MiniPrep, CavroSterile and PCR Ready
904-011G1000 μLFilteredTecan Genesis, Tecan Freedom EVO, MiniPrep, CavroSterile and PCR Ready
904-252
also known as 904-252GP
1000 μLNon-filteredCavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusCertified Quality
901-01110 μLFilteredTecan Genesis, Tecan Freedom EVO, MiniPrep, CavroSterile and PCR Ready
Catalog number 904-253
also known as 904-253GPA
Price (SGD)
1,062.00
CS
Add to cart
Volume (Metric):
1000 μL
Filtered:
Non-filtered
For Use With (Equipment):
Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for Purpose:
Certified Quality
Price (SGD)
1,062.00
CS
Add to cart
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
Features:
  • Available as sterile, filtered and/or liquid sensing.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlack
AutoclavableAutoclavable
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
FormatRack
Tip StyleConductive
Quantity4800 Tips
For Use With (Equipment)Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for PurposeCertified Quality
Unit SizeCS