Accessories for Digital Mini Rotators
Accessories for Digital Mini Rotators
Actual product may vary
Accessories for Digital Mini Rotators
Get assorted accessories for use with Digital Mini Rotators
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDimensions (L x W)
88882119Tube support 50 mL262 x 112 mm
88882112Sticky Mat, 14 x 14 cm140 x 140 mm
88882113Flat Platform w/ Rubber Mat330 x 270 mm
88882104Universal Attachment307 x 303 mm
Catalog number 88882119
Price (SGD)
174.00
EA
Add to cart
Description:
Tube support 50 mL
Dimensions (L x W):
262 x 112 mm
Price (SGD)
174.00
EA
Add to cart
Assorted accessories for use with Digital Mini Rotators
Specifications
Tube Capacity50 mL
Product TypeTube Support, 50 mL
MaterialPlastic
Dimensions (L x W)262 x 112 mm
Dimensions (L x W x H)262 x 112 x 140 mm
DescriptionTube support 50 mL
For Use With (Equipment)Rocker, Rotator, Mixer
Unit SizeEA