Digital Tube Rockers
Digital Tube Rockers
Actual product may vary
Digital Tube Rockers
Digital Tube Rockers
Digital Tube Rockers
Digital Tube Rockers
Digital Tube Rockers
Digital Tube Rockers
Digital Tube Rockers
Digital Tube Rocker is a precision-controlled platform mixer that accommodates double capacity in a compact footprint for smooth rocking and uniform suspension.
Have Questions?
Catalog NumberPlug TypeCertifications/Compliance
88882018EU, UK, CN PlugCE
Catalog number 88882018
Price (SGD)
640.00
Special offer
800.00
Save 160.00 (20%)
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Certifications/Compliance:
CE
Price (SGD)
640.00
Special offer
800.00
Save 160.00 (20%)
EA
Add to cart
The Thermo Scientific™ Digital Tube Rocker provides smooth agitation for mixing samples in test tubes. Designed for uniform specimen suspension with a compact footprint.
  • Digital display of Speed and Tilt angle
  • Speed range: 2 – 40 rpm
  • Speed Accuracy: ±1 rpm
  • Adjustable rocking Angle: 1-48° (±24°)
  • Angle Accuracy: ±1°
  • Tube Capacity:
    • Standard: 16 x 10-20mm tube / 7 x 22-30mm tube
    • Double Platform: 30 x10-20mm tube /14 x 22-30mm tube
  • Optional double platform offers big tube capacity.
Specifications
Speed Control Accuracy±1 rpm
Speed Range2 to 40 rpm
Electrical Requirements100/240 V
Amperage0.1 A
HoldsTubes
Tube Size30 mm dia.
Product TypeDigital Tube Rocker
Certifications/ComplianceCE
Plug TypeEU, UK, CN Plug
Frequency50/60 Hz
MaterialMetal
Dimensions (D x W x H) Interior411x144x146 mm (16.2 x 5.7 x 5.7 in.)
Dimensions (L x W x H)330 x 140 x 12 mm (13 x 5.5 x 0.5 in.)
Shipping Weight (English)9.9 lb.
Shipping Weight (Metric)4.5 kg
Weight (English)7.5 lb.
Weight (Metric)3.4 kg
Controller TypeDigital Control
ModelDigital Tube Rocker
DisplayDigital
For Use With (Equipment)88882158, 88882159, 88882160, diameter 5mm - 30 mm tubes
Humidity Range20 to 85%
IncludesDimple rubber mat 88882159
Motor TypeDC Motor
No. of Positions14
Platform StyleTray
Unit SizeEA