Digital Vortex Mixers
Digital Vortex Mixers
Actual product may vary
Digital Vortex Mixers
Digital Vortex Mixers
Digital Vortex Mixers
Digital Vortex Mixers
Get accurate and efficient sample mixing
Have Questions?
Catalog NumberPlug TypeVoltage
88882010EU, UK, CN Plug230 V
Catalog number 88882010
HSN 84798200
Price (SGD)
463.00
Special offer
Ends: 31-Mar-2026
464.00
Save 1.00
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Voltage:
230 V
Price (SGD)
463.00
Special offer
Ends: 31-Mar-2026
464.00
Save 1.00
EA
Add to cart
Designed for accurate speed control for continuous mixing or pulsing of samples
  • Digital display of Speed and Time
  • Continuous and Touch modes
  • Speed range: 200 to 3000 rpm
  • Precise speed control: ±5% rpm
  • Quick set up to Max rpm:
  • 3 seconds 1500 rpm (Touch)
  • 6 seconds 3000 rpm (Continuous)
  • Time set: up to 99 minutes
  • 2 Timer modes:
    • Down-counting for repeat tests
    • Timer: Continuous, Time set as 0
  • Various accessories for different vessels
Specifications
Load Bearing Capacity (Metric)0.5 kg
Speed Range200 to 3000 rpm
Electrical Requirements100/240 V
Voltage230 V
Product TypeDigital Vortex Mixer
Certifications/CompliancecCSAus, CE, Certificate of Analysis (COA), Certificate of Analysis (COA)
Plug TypeEU, UK, CN Plug
Frequency50/60 Hz
MaterialRubber, Plastic Platform
Dimensions (L x W x H)8.1 x 5.9 x 5.2 in. (20.5 x 15.1 x 13.1 cm)
Height (English)5.2 in.
Height (Metric)13.1 cm
Length (English)8.1 in.
Length (Metric)20.5 cm
Width (English)5.9 in.
Width (Metric)15.1 cm
Shipping Weight (English)14.6 lb.
Shipping Weight (Metric)6.6 kg
ModelDigital Vortex Mixer
For Use With (Equipment)Rocker, Rotator, mixer
Operating ModesTouch, Continuous
Timer Range1 to 99 min., Continuous
Warranty and Service Offering2 Years
Unit SizeEA