CO<sub>2</sub> Resistant Shakers
Actual product may vary
CO2 Resistant Shakers
Benefit from reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug TypeIncludes
88881104EU, UK, CN PlugShaker, Universal Platform, Flask Clamp Starter Kit
88881102EU, UK, CN PlugShaker, Control Box
Catalog number 88881104
HSN 84798200
Price (SGD)
4,917.00
Special offer
Ends: 30-Jun-2026
5,784.00
Save 867.00 (15%)
Each
Plug Type:
EU, UK, CN Plug
Includes:
Shaker, Universal Platform, Flask Clamp Starter Kit
Price (SGD)
4,917.00
Special offer
Ends: 30-Jun-2026
5,784.00
Save 867.00 (15%)
Each
Get reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment with Thermo Scientific™ CO2 Resistant Shakers.
  • Orbital shaking through magnetic power eliminates the need to replace belts
  • Small footprint ideal for use in a CO2 incubator
  • Specially treated mechanical components protect up to 20% CO2 and 95% humidity conditions
  • Convenient external control box with LED display allows for easy adjustments without opening the chamber door
  • Minimal heat dissipation assures conditions within the CO2 chamber are not effected
  • Stainless steel construction for protection from corrosive CO2 gassing conditions
  • An extensive selection of accessories fits most applications
  • CO2 resistant shaker can be used outside of CO2 incubator for maximum value
  • External control box for ease-of-use without disrupting your samples
A large selection of available accessories to fit most applications:
  • Wide variety of flask clamps available
  • Fixed and adjustable tube racks available
  • 96-well microplate holder
  • Rubber and sticky mats
  • Flask clamp starter kit for ease and flexibility
Applications:
  • Mammalian cell culture
  • Bacterial suspensions
  • Staining and destaining
  • Washing procedures
  • Hybridization
  • Solubility studies
  • Extraction procedures
Warranty:
  • 2 year Thermo Fisher Scientific standard limited warranty. See your sales representative for details.
Specifications
Orbit19 mm
Load Bearing Capacity (Metric)6 kg
Speed Control Accuracy1 rpm
Speed Range30 to 300 rpm
Electrical Requirements100/240 V, 50/60 Hz
Voltage100/240 V
Temperature Range (Metric)4°C to 60°C
Load Bearing Capacity (English)13 lb.
Certifications/CompliancecCSAus, CE Mark, RoHS, WEEE
Plug TypeEU, UK, CN Plug
Frequency50/60 Hz
Dimensions (L x W x H)11.8 x 14.2 x 3.8 in. (300 x 360 x 96 mm)
Weight (English)29 lb.
Weight (Metric)13 kg
Controller TypeCo2-Resistant
ModelCO2 Resistant Shaker
DisplayLED, speed and time
IncludesShaker, Universal Platform, Flask Clamp Starter Kit
Timer Range0 to 99:59 hr., Continuous
Unit SizeEach