Bottle/Tube Roller
Bottle/Tube Roller
Actual product may vary
Bottle/Tube Roller
Safely use this sleek, compact bottle/tube roller, capable of rolling multiple size tubes or bottles, in CO2 and/or humid environments.
Have Questions?
Catalog NumberSpeed RangeTemperature Range (Metric)Electrical RequirementsPlug Type
888810041 to 80 rpm4°C to 60°C100/240 V, 50/60 HzCord set with various plugs
Catalog number 88881004
Price (SGD)
2,151.00
EA
Add to cart
Speed Range:
1 to 80 rpm
Temperature Range (Metric):
4°C to 60°C
Electrical Requirements:
100/240 V, 50/60 Hz
Plug Type:
Cord set with various plugs
Price (SGD)
2,151.00
EA
Add to cart
Safely use the sleek, compact Thermo Scientific™ Bottle/Tube Roller in CO2 and/or humid environments. The Bottle/Tube Roller is capable of rolling multiple size tubes or bottles and features digital controlled speed ranging from 1- 80rpm. Maximize valuable bench or incubator space by stacking two high with the optional stacking kit. Control without disturbing incubating samples with the optional remote control, which also adds a timer function when used. Includes O Rings to ensure smooth rolling when using tubes.
  • Adjustable speed accommodates a variety of applications
  • Anti-corrosive, wear-resistant construction enables use in humid and CO2 environments
  • Adjustable rollers accommodate a variety of tube/bottle sizes
  • Stacking kit allows stacking two units, saving valuable bench space
  • Optional remote control facilitates operating unit while in incubator and adds timer function
Includes:
O Rings to ensure smooth rolling when using tubes
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Load Bearing Capacity (Metric)6.5 kg
Speed Range1 to 80 rpm
Electrical Requirements100/240 V, 50/60 Hz
Voltage100/240 V
Temperature Range (Metric)4°C to 60°C
Wattage10 W
Product TypeRollers
Load Bearing Capacity (English)14.3 lb.
Certifications/CompliancecCSAus, CE
Plug TypeCord set with various plugs
Temperature Range (English)39.2°F to 140°F
Frequency50/60 Hz
Diameter (English) Roller1.1 in.
Max. Bottle Diameter (English)≤ 4.72 in.
Diameter (Metric) Roller28 mm
Max. Bottle Diameter (Metric)≤ 120 mm
Dimensions (L x W x H)14.2 x 2.8 in. (360 x 72 mm)
Height (English)2.8 in.
Height (Metric)72 mm
Length (English)14.2 in.
Length (Metric)360 mm
Width (English)14.9 in.
Width (Metric)378 mm
Shipping Weight (English)14.3 lb.
Shipping Weight (Metric)6.5 kg
ModelBottle, Tube Roller
DisplayLED Roller Speed Display
Humidity Range≤95%
No. of Rollers6 rollers standard, up to 11 can be added
StackableTwo high with optional kit
Timer Range0 to 999 min., Continuous
Tolerance≤ 95% humidity and CO2 resistant
Warranty and Service Offering2 Years
Unit SizeEA