Search Thermo Fisher Scientific
Catalog Number | No. of Places | Tube Size |
---|---|---|
88880136 | 24 | 13mm |
88880129 | 48 | 0.5mL |
88880130 | 30 | 1.5mL |
88880131 | 15 | 15mL |
88880132 | 6 | 50mL small |
88880133 | 6 | 50mL large |
Share catalog number, name or link