RT Basic Series Magnetic Stirrers
RT Basic Series Magnetic Stirrers
Actual product may vary
RT Basic Series Magnetic Stirrers
Experience the reliability of our plug-and-play magnetic stirrers, ideal for routine stirring applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityTop Plate Diameter (Metric)Top Plate Diameter (English)Plug Type
888800104 L170 mm6.69 in.Cord set w/various plugs
888800082 L120 mm4.72 in.Cord set w/various plugs
Catalog number 88880010
Price (SGD)
485.00
EA
Add to cart
Capacity:
4 L
Top Plate Diameter (Metric):
170 mm
Top Plate Diameter (English):
6.69 in.
Plug Type:
Cord set w/various plugs
Price (SGD)
485.00
EA
Add to cart
Experience reliability with the plug-and-play Thermo Scientific™ RT Basic Series Magnetic Stirrers, ideal for routine stirring applications. Three size options to choose from — 120mm, 170mm, or 220mm — all with a low-profile, lightweight design that has an extremely small footprint. The RT basic stirrers feature a chemical-resistant polypropylene top and body, a strong magnetic coupling to ensure consistent stirring during experimental procedures, and a brushless DC Motor that will not generate heat. Analog speed control ranges from 150 to 2500rpm. All units include two non-slip silicone plate covers for added protection from spills.
  • Low-profile, lightweight design with small footprint
  • Strong magnetic coupling to ensure consistent stirring during experimental procedures
  • Brushless DC Motor will not generate heat and “cook” sample
  • Polypropylene top and body provides excellent chemical resistance 
  • Three size options — 120mm, 170mm, or 220mm 
  • Analog speed control: 150 to 2500rpm
  • Detachable plug
  • Two non-slip silicone plate covers provided with all units (1 black, 1 white)
  • Low power consumption 
  • 2 year warranty
Specifications
Speed150 to 2500 rpm
Top Plate MaterialPolypropylene
Capacity4 L
ShapeRound
Voltage100/240 V
Wattage6 w
Dimensions (L x W x H)7.28 x 7.28 x 2.56 in. (185 x 185 x 65 mm)
Height (English)2.56 in.
Height (Metric)65 mm
Length (English)7.28 in.
Length (Metric)185 mm
Top Plate Diameter (English)6.69 in.
Top Plate Diameter (Metric)170 mm
Width (English)7.28 in.
Width (Metric)185 mm
Electrical Requirements100 to 240V 50/60Hz
Frequency50/60 Hz
Plug TypeCord set w/various plugs
Certifications/ComplianceCE
Motor TypeBrushless DC
TypeStirrer
Unit SizeEA