Block, 1.5 mL and 2.0 mL
Block, 1.5 mL and 2.0 mL
Actual product may vary
Block, 1.5 mL and 2.0 mL
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog NumberNo. of Places
8887011318
Catalog number 88870113
Price (SGD)
259.00
EA
Add to cart
No. of Places:
18
Price (SGD)
259.00
EA
Add to cart
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
DescriptionMixed block, 1.5mL and 2.0mL
Capacity (Metric)18 x 1.5 mL, 10 x 2 mL
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds18 x 1.5mL and 10 x 2.0mL
LightingLED
No. of Places18
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
Unit SizeEA