Block, 40 x 0.5 mL
Block, 40 x 0.5 mL
Actual product may vary
Block, 40 x 0.5 mL
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog number 88870110
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
Description40 x 0.5mL block
Capacity (Metric)0.5 mL
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds40 x 0.5mL
LightingLED
No. of Places40
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
Unit SizeEA