18/10 Stainless Steel Round Bottom Bowl
18/10 Stainless Steel Round Bottom Bowl
Actual product may vary
18/10 Stainless Steel Round Bottom Bowl
Have Questions?
Catalog NumberCapacity (Metric)
878050 mL
Catalog number 8780
Price (SGD)
44.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
44.00
PK
Add to cart
Specifications
Bottom ShapeRound
Capacity (Metric)50 mL
MaterialStainless Steel
ShapeRound
TypeBowl
Height (Metric)30 mm
Diameter (Metric)60 mm
Unit SizePK