Thermo Scientific™ QSP™ Microtiter Tube Plug Strip Caps
Product Image
Thermo Scientific™ QSP™ Microtiter Tube Plug Strip Caps
Thermo Scientific™ QSP™ Plug Strips (polyethylene LDPE) seal tightly for storage. With eight plugs per strip, they may be used with rows of tubes or easily cut apart for individual samples. The plug strips are available pre-sterilized as the material will not withstand autoclaving.
Have Questions?
Catalog NumberAutoclavableMaterial
845-QAutoclavablePolypropylene
Catalog number 845-Q
Price (SGD)
42.00
EA
Add to cart
Autoclavable:
Autoclavable
Material:
Polypropylene
Price (SGD)
42.00
EA
Add to cart
Plug Strips (polyethylene LDPE) seal tightly for storage. With eight plugs per strip, they may be used with rows of tubes or easily cut apart for individual samples. The plug strips are available pre-sterilized as the material will not withstand autoclaving.
  • 1.2ml volume capacity tube
  • Packaging available in bulk, rack or rack sterile options
  • 8 plug strip tubes used to seal tubes, available sterile and non sterile
Specifications
MaterialPolypropylene
Volume (Metric)1.2 mL
Product LineQSP
Volume (Metric) Total1.2 mL
AutoclavableAutoclavable
TypeMicrotiter Cluster Tube
Unit SizeEA