Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Have Questions?
Catalog Number
Type
840-278000
Thermal Snap-on Printer
Catalog number 840-278000
Price (SGD)
-
Type:
Thermal Snap-on Printer
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible In addition to valuable built-in features such as the high resolution color display, large sample compartment, and data export options, a complete selection of accessories are available for the the GENESYS 30 spectrophotometer to help meet the needs of students, educators and lab technicians for accurate quantitative analysis.
Holds one rectangular longpath cell 20 - 100mm pathlength
Comes assembled in a removable sample compartment liner
Allows for flexibility between samples
Specifications
TypeThermal Snap-on Printer
For Use With (Application)Direct hard-copy printing of results from the instrument onboard software control.
For Use With (Equipment)GENESYS 30 Visible Spectrophotometer