Nalgene™ Polypropylene, Carboy with Spigot
Nalgene™ Polypropylene, Carboy with Spigot
Actual product may vary
Nalgene™ Polypropylene, Carboy with Spigot
For Use With: Long-term storage, and dispensing solutions and media; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Catalog number 8319-0020
Price (SGD)
195.00
EA
Add to cart
Price (SGD)
195.00
EA
Add to cart
For Use With: Long-term storage, and dispensing solutions and media; Autoclavable; Closure Material: Polypropylene; Color: White
Features:
  • Molded in graduations in 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles make lifting comfortable
  • Ideal for storing and dispensing solutions and media and can be autoclaved before refilling to eliminate bacterial growth
  • Molded in spigot boss provides strength and leak resistance
  • Autoclavable/Graduated/Leakproof†
Includes:
Polypropylene screw cap
Note:
  • Containers can be used for long-term storage, but direct UV exposure should be avoided
  • Neck accepts No. 13.5 rubber stopper
  • Spigot closure included.
  • Follow product insert instructions for autoclaving to ensure proper function
  • Tubing adaptor is not autoclavable
.Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com
Specifications
AutoclavableNot Autoclavable
Capacity (English)2.6 gal.
Capacity (Metric)10 L
Closure MaterialPolypropylene
Closure Size83B
ColorWhite
DescriptionAutocavable Carboy w/Spigot
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer250 mm
For Use With (Application)Long-term storage, and dispensing solutions and media
GraduationsMolded-in graduations in 1 gal. and 5L increments
Height (Metric) With Closure389 mm
IncludesIntegral shoulder handles, molded-in spigot boss
MaterialPolypropylene
No. per Case6/Cs.
No. per Pack1 Ea.
Product TypeCarboy with Spigot
Unit SizeEA