Nalgene™ LDPE, Round Carboy with Spigot
Nalgene™ LDPE, Round Carboy with Spigot
Actual product may vary
Nalgene™ LDPE, Round Carboy with Spigot
For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Have Questions?
Catalog number 8318-0130
Price (SGD)
346.00
EA
Add to cart
Price (SGD)
346.00
EA
Add to cart
For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Features:
  • Molded in graduations of 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles that make lifting comfortable for four largest sizes
  • Molded-in spigot boss for strength and leak resistance
  • Leakproof†
Ordering Information:
Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.
Includes:
Polypropylene spigot and screw closure
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.
Specifications
AutoclavableNot Autoclavable
Capacity (English)13 gal.
Capacity (Metric)50 L
Closure MaterialPolypropylene
Closure Size83B
ColorWhite
DescriptionRound Carboys w/Spigot
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer379 mm
For Use With (Application)Collect and dispense distilled water, acids and reagents
GraduationsMolded-in graduations in 1 gal. and 5L increments
Height (Metric) With Closure678 mm
IncludesIntegral shoulder handles, molded-in spigot boss
MaterialLDPE
No. per Case1 Ea.
No. per Pack1 Ea.
Product TypeCarboy with Spigot
Unit SizeEA