Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Actual product may vary
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers
Store samples confidently in the Forma 8600 Series -86°C chest freezer. Available in four sizes to accommodate a variety of needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsHoldsDimensions (DxWxH) ExteriorPlug Type
820CV230 V 50 HzBoxes (396)33.4 x 96 x 40.5 in.EU
803CV230 V 50 HzBoxes (63)29.5 x 28.5 x 43.8 in.EU
817CV230 V 50 HzBoxes (348)33.8 x 87.6 x 40.5 in.EU
Catalog number 820CV
HSN 84183090
Price (SGD)
-
Electrical Requirements:
230 V 50 Hz
Holds:
Boxes (396)
Dimensions (DxWxH) Exterior:
33.4 x 96 x 40.5 in.
Plug Type:
EU
Store samples confidently in Thermo Scientific™ Forma 8600 Series -86°C ultra-low temperature chest freezers. Available in four sizes, the 8600 series accommodates a variety of needs and delivers all the same sample protection features as our Forma 900 and Forma 7000 upright series.
Key Features:
  • Centralized information center, including microprocessor control and monitoring system, ensures all controls and displays are easy to reach and read—knee level on the 3 cu. ft. models.
  • Freezer improves temperature control and increases reserve BTUH capacity, resulting in a more stable temperature sample.
  • Polystyrene interior sub-lids reduce cold air loss and improve temperature recovery after door openings.
  • Easy-to-remove, washable filter provides protection from dust on the condenser that causes reduced refrigeration performance and increased risk to samples.
  • Easy-to-use ergonomic door handle offers single-hand operation.
  • Choice of CO2 or LN2 safety back-up systems offer additional sample protection in the event of a power or mechanical failure.
Construction:
  • 5 in. (127mm) foamed-in-place, polyurethane insulation reduces power consumption and maintains temperature setpoint.
  • Triple-point door gasket provides longer holding time in the event of a door opening.
  • Rugged, heavy gauge, cold-rolled steel cabinets are finished with powder-coat paint for a uniform exterior that resists chipping and rust.
  • 1 in. (25mm) access port allows for the use of inexpedient probes or instrumentation.
  • Interior is stainless steel.
Warranty varies by country, please contact us for details
Specifications
DescriptionForma 800 Series Ultra-Low Chest Freezer, 396-Box Capacity, 230V, 50Hz
TypeUltra Low Freezer, Chest
Capacity (English)20 cu. ft.
Capacity (Metric)566.3 L
Temperature Range-50°C to -86°C
Cabinet MaterialCold-rolled Steel
InsulationPolyurethane Foam
InteriorStainless Steel
Certifications/ComplianceCE, cULus, FDA Class I, UL
StyleChest
Condenser TypeFin and Tube, Forced-Air Cooled
Control TypePush Button LED
Product LineForma
WarrantyWarranty varies by country, please contact us for details
DefrostManual
ModelForma
Water CooledOptional
Dimensions (DxWxH) Exterior33.4 x 96 x 40.5 in.
Dimensions (D x W x H) Interior18.5 x 66.5 x 28 in. (47 x 168.9 x 71.1 cm)
Monitoring OptionsAlarm, Recorder
Access SecurityKey Lock
Chart RecorderOptional 7-day chart recorder
Data OutputsRS-485, 4-20mA, dry contacts - standard
Operating ModesStandard Operation
Setpoint SecuritySingle ID
Backup SystemsCO2 or LN2 Optional
Smart View CompatibilitySmart-Vue Compatible
Amperage12 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Wattage970 W
Voltage CompensationBuck/Boost
Vial Capacity39,600 x 2 mL
CompressorHermetic
RefrigerantR404A (1st stage), R508B (2nd stage)
Shipping Weight (English)833 lb.
Shipping Weight (Metric)378 kg
Weight (English)726 lb.
Weight (Metric)363 kg
HoldsBoxes (396)
Unit SizeEach
Frequently asked questions (FAQs)
The low battery alarm is displaying on my ULT (Ultra-Low Temperature) freezer. Can I buy a new battery?
You can order a new battery (Cat. No. 400159) online.