Search Thermo Fisher Scientific
Catalog Number | Holds | Dimensions (L x W x H) |
---|---|---|
820109 | 100 Cryo Tubes | 5 x 5 x 2 in. |
FIS11-678-24A | Cryo Tubes | 5 x 5 x 2 in. |
820002 | 81 Cryo Tubes | 5 x 5 x 2 in. |
FIS13-994-070 | 81 Cryo Tubes | 5 x 5 x 2 in. |
FIS13-994-071 | 81 Cryo Tubes | 5 x 5 x 3 in. |
FIS82-010-9 | 100 Cryo Tubes | 5 x 5 x 2 in. |
Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.