Matrix™ Reagent Reservoirs
Actual product may vary
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MaterialPackagingType
809425 mLPolystyrene50 Individually wrappedReagent Reservoir
FKS1049597375 mLPolypropylene10 pcs /bagReservoir Lid
8085100 mLPolystyrene10 Bags of 10Reagent Reservoir
FIS14-387-067100 mLPolystyrene10 Bags of 10Reagent Reservoir
FKS10141873100 mLPolystyrene10 Bags of 10Reagent Reservoir
8086100 mLPolystyrene50 Individually wrappedReagent Reservoir
FIS14-387-068100 mLPolystyrene50 Individually wrappedReagent Reservoir
FKS10375023100 mLPolystyrene50 Individually wrappedReagent Reservoir
FIS14-387-06925 mLPolystyrene10 Bags of 10Reagent Reservoir
FKS1075635425 mLPolystyrene10 Bags of 10Reagent Reservoir
809525 mLPolystyrene10 Bags of 10Reagent Reservoir with Divider
FIS14-387-07125 mLPolystyrene10 Bags of 10Reagent Reservoir with Divider
FKS1022574325 mLPolystyrene10 Bags of 10Reagent Reservoir with Divider
FIS14-387-07225 mLPolystyrene50 Individually wrappedReagent Reservoir with Divider
Catalog number 8094
HSN 39269099
Price (SGD)
171.00
CS
Capacity (Metric):
25 mL
Material:
Polystyrene
Packaging:
50 Individually wrapped
Type:
Reagent Reservoir
Price (SGD)
171.00
CS
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Specifications
ColorWhite
Capacity (Metric)25 mL
MaterialPolystyrene
SterilitySterile
TypeReagent Reservoir
Packaging50 Individually wrapped
Unit SizeCS