Blood Adapter
Blood Adapter
Actual product may vary
Blood Adapter
Blood Adapter for triple/quad blood bags. For the H-12000 Rotor.
Have Questions?
Catalog NumberFor Use With (Equipment)
77030450 to 500 mL Triple/Quad Blood Bag, H-12000 Rotor
Catalog number 77030
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
450 to 500 mL Triple/Quad Blood Bag, H-12000 Rotor
  • Blood Adapter for triple/quad blood bags.
  • For the H-12000 Rotor.
Specifications
TypeBlood Bag Adapter
MaterialPlastic
Capacity450 to 500 mL
AutoclavableNot Autoclavable
For Use With (Equipment)450 to 500 mL Triple/Quad Blood Bag, H-12000 Rotor
Unit SizeEA