For Use With: For Water Jacket models, Steri-cycle and direct heat models (31X, 32X, 37X & 38X) and 8000 Series Incubators
Have Questions?
Catalog Number
Type
760175
HEPA Filter
Catalog number 760175
Price (SGD)
151.00
EA
Add to cart
Type:
HEPA Filter
Price (SGD)
151.00
EA
Add to cart
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
99.97% filtering at 0.3μm
Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Specifications
DescriptionReplacement HEPA Filter
For Use With (Equipment)Forma™ Steri-Cycle™, Direct Heat and Water Jacket