General Purpose Centrifuge Rotor Packages
Actual product may vary
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Have Questions?
Catalog NumberModel
75016107TX-400 Blood Tube Package
Catalog number 75016107
Price (SGD)
2,352.00
Special offer
Ends: 30-Nov-2026
4,704.00
Save 2,352.00 (50%)
Each
Model:
TX-400 Blood Tube Package
Price (SGD)
2,352.00
Special offer
Ends: 30-Nov-2026
4,704.00
Save 2,352.00 (50%)
Each
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
MaterialSee individual centrifuge, rotor and accessory data
TypeRotor Equipment Package
CapacityMax in TX-400: 76 x 5/7 mL Blood Tubes, 56 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net WeightSee individual components of package
For Use With (Equipment)Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
IncludesTX-400 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
WarrantySee Manual
BiocontainmentYes
ModelTX-400 Blood Tube Package
Unit SizeEach