Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
Actual product may vary
Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorTube Size
75008144
also known as 75008144FS
White175/250 mL
75008137
also known as 75008137FS
Olive Brown15 mL
Catalog number 75008144
also known as 75008144FS
Price (SGD)
831.00
EA
Add to cart
Color:
White
Tube Size:
175/250 mL
Price (SGD)
831.00
EA
Add to cart
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
For Use With (Equipment)LH-4000W and LH-4000 rotor with round bucket
ColorWhite
Tube Size175/250 mL
Unit SizeEA