Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
Actual product may vary
Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Have Questions?
Catalog NumberTube TypeTube Size
75007621Conical15 mL
Catalog number 75007621
Price (SGD)
217.00
EA
Add to cart
Tube Type:
Conical
Tube Size:
15 mL
Price (SGD)
217.00
EA
Add to cart
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Tube TypeConical
For Use With (Equipment)4-place Swing rotor with round bucket
Tube Size15 mL
Unit SizeEA