Adapters for TX-1000 Swinging Bucket Rotor
Actual product may vary
Thermo Scientific
Adapters for TX-1000 Swinging Bucket Rotor
An assortment of adapters for outstanding flexibility
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Equipment)
750073058 x 250 mL250mL Thermo Scientific Nalgene Bottle
75003697100 x 9/10 mL9/10mL Blood Collection Tube
75003709164 x 4.5/6 mL4.5/6mL Blood Collection Tube
75003829150 to 300 mL Small Blood BagsSmall bag/cell culture bags 4 x 2 bags (< 350mL)
750073014 x 1000 mL1000mL Bio-Bottle
750042534 x 500 mL500mL Nalgene Bottle
750053924 x 250 mL250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
75007302500 mL500mL Corning Bottle
750073034 x 6 mL Standard or 4 x 2 mL Deep-WellMicroplate carriers
750073044 x 750 mL750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
7500730696 x 15 mL15mL Conical Tube
Catalog number 75007305
HSN 84219100
Price (SGD)
1,396.00
Each
Capacity:
8 x 250 mL
For Use With (Equipment):
250mL Thermo Scientific Nalgene Bottle
Price (SGD)
1,396.00
Each
An assortment of adapters for outstanding flexibility
Specifications
TypeRotor Adapter
Tube Type250 mL Bottle
Capacity8 x 250 mL
Max. Tube Diameter62 mm
Max. Tube Length145 mm
For Use With (Equipment)250mL Thermo Scientific Nalgene Bottle
Unit SizeEach