Check Order Status
Centrifuge Rotor Adapters
Actual product may vary
Centrifuge Rotor Adapters
Have Questions?
Catalog Number
For Use With (Equipment)
75006533
also known as 75006533FS
Kendro centrifuge rotor
Catalog number
75006533
also known as 75006533FS
Price (SGD)
277.00
EA
Add to cart
For Use With (Equipment):
Kendro centrifuge rotor
Price (SGD)
277.00
EA
Add to cart
Specifications
For Use With (Equipment)
Kendro centrifuge rotor
Color
Green