Rotor Packages for Sorvall™ ST8 Benchtop Centrifuge
Actual product may vary
Rotor Packages for Sorvall™ ST8 Benchtop Centrifuge
Rotor Packages for Sorvall™ ST8 Benchtop Centrifuge
Rotor Packages for Sorvall™ ST8 Benchtop Centrifuge
Convenient cell culture and clinical packages
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityIncludes
7500576224 x 0.7 mLTX-150 Rotor (75005701), Set of 4 Round Buckets (75005702), Set of 4 ClickSeal Biocontainment Lids (75005707), Sets of 4 Adapters for Blood Collection Tubes: 5/7mL (75005739) and 10mL (75005738)
750057604 x 50 mLTX-150 Rotor (75005701), Set of 4 Round Buckets (75005702), Set of 4 Adapters for 50mL Conical Tubes (75005736)
750057618 x 50 mLTX-150 Rotor (75005701), Set of 4 Conical Buckets (75005703), Set of 2 Adapters for 15mL Conical Tubes (75005808)
Catalog number 75005762
HSN 84219100
Price (SGD)
4,092.00
Each
Capacity:
24 x 0.7 mL
Includes:
TX-150 Rotor (75005701), Set of 4 Round Buckets (75005702), Set of 4 ClickSeal Biocontainment Lids (75005707), Sets of 4 Adapters for Blood Collection Tubes: 5/7mL (75005739) and 10mL (75005738)
Price (SGD)
4,092.00
Each
Convenient cell culture and clinical packages
Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to certified Thermo Scientific™ ClickSeal™ biocontainment lids—shorten run set-up time while providing peace-of-mind that your samples are secure.
Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers:
  • Trouble-free rotor installation and removal:
    • No tools are required
    • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
  • Biocontainment sealing, including certified ClickSeal lids for glove-friendly, one-handed operation
  • 8 adapters available to meet your evolving clinical and research needs
Specifications
MaterialAluminum
TypeSwinging Bucket Rotor
Capacity24 x 0.7 mL
Max. RCF3,260 x g
Max. Speed4,500 rpm
Radius (Metric)5.1 to 14.4 cm
Max. Tube Length12.4 cm
Max. Tube Diameter1.8 cm
For Use With (Equipment)Thermo Scientific™ Sorvall™ ST8 Benchtop Centrifuge
Angle90°
IncludesTX-150 Rotor (75005701), Set of 4 Round Buckets (75005702), Set of 4 ClickSeal Biocontainment Lids (75005707), Sets of 4 Adapters for Blood Collection Tubes: 5/7mL (75005739) and 10mL (75005738)
BiocontainmentYes
Unit SizeEach