Check Order Status
Adapters for Thermo Scientific™ MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Adapters for Thermo Scientific™ MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
PCR Tube and Microtube Adapters
Have Questions?
Contact Us
Change view
Catalog Number
Capacity
Tube Type
75005753
0.5 mL
Microtube
75005752
0.2 mL
PCR
2 Options
Catalog number
75005753
Price (SGD)
531.00
EA
Estimated availability date
23-Nov-2025
Add to cart
Capacity:
0.5 mL
Tube Type:
Microtube
Price (SGD)
531.00
EA
Add to cart
Adapters for Thermo Scientific MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
Specifications
Type
Rotor Adapter
Tube Type
Microtube
Capacity
0.5 mL
For Use With (Equipment)
MicroClick 30 x 2 and MicroClick 24 x 2 rotor
Tube Size
0.5 mL
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link