Adapters for TX-150 Swinging Bucket Rotors
Adapters for TX-150 Swinging Bucket Rotors
Actual product may vary
Adapters for TX-150 Swinging Bucket Rotors
Assorted tube adapters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube Type
7500573650 mL Conical or Skirted Tube
7500574425 mL Universal Flat/Skirted Tube
750057403/5 mL Blood Collection Tube
7500573711 mL IVF Tube
750057395/7 mL Blood Collection Tube
7500350413 mL Urine Tube
Catalog number 75005736
Price (SGD)
704.00
EA
Add to cart
Tube Type:
50 mL Conical or Skirted Tube
Price (SGD)
704.00
EA
Add to cart
Assorted tube adapters
Specifications
TypeRotor Adapter
Tube Type50 mL Conical or Skirted Tube
Capacity50 mL
Max. Tube Diameter29.5 mm
Max. Tube Length120 mm
For Use With (Equipment)50mL Conical Buckets for TX-150 rotor
Unit SizeEA