TX-100 Swinging Bucket Clinical Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
TX-100 Swinging Bucket Clinical Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Actual product may vary
TX-100 Swinging Bucket Clinical Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
TX-100 Swinging Bucket Clinical Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
TX-100 Swinging Bucket Clinical Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Efficiently process up to 16 x 5/7mL blood collection tubes per run.
Have Questions?
Catalog NumberCapacity
7500570516 x 15 mL
Catalog number 75005705
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
16 x 15 mL
Efficiently process up to 16 x 5/7mL blood collection tubes per run with the Thermo Scientific™ TX-100 Swinging Bucket Clinical Rotor.
Ordering Information:
Includes unsealed carriers. Adapters are sold separately. Contact your sales representative for more information.
Specifications
DescriptionTX-100 Swinging Bucket Clinical Rotor
TypeSwinging Bucket Rotor
Capacity16 x 15 mL
Max. RCF3,260 x g
Max. Speed4,500 rpm
Radius (Metric)144 mm
K Factor4258
Max. Tube Length121 mm
Max. Tube Diameter17 mm
For Use With (Equipment)Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle90°
Unit SizeEA