Sorvall™ ST 16 Centrifuge Series
Sorvall™ ST 16 Centrifuge Series
Actual product may vary
Sorvall™ ST 16 Centrifuge Series
Perform cell culture and blood processing applications using the Thermo Scientific™ Sorvall™ ST 16 Centrifuge Series, which is compatible with a wide range of rotors.
Have Questions?
Catalog NumberElectrical RequirementsDescription
75004240230 V 50/60 HzSorvall ST 16 Centrifuge
Catalog number 75004240
Price (SGD)
-
Electrical Requirements:
230 V 50/60 Hz
Description:
Sorvall ST 16 Centrifuge
Perform cell culture and blood processing applications using the Thermo Scientific™ Sorvall™ ST 16 Centrifuge Series, which is compatible with a wide range of rotors.
Easy To Use
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
Auto-Lock™ III Rotor System
  • Secure locking system allows easy push-button installation and exchange of rotors
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning, a healthier working environment and longer unit life
ClickSeal™ Bucket Sealing System
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR™ in Porton Down, UK
SMARTSpin™ Technology
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
Space-Saving Design
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning
Outstanding Energy Efficiency
  • Up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing
Ventilated Model ST 16
  • Dimensions: 23.8L × 17.3W × 14.2 in.H [34.3 in.H open] (60.5 × 44 × 36cm [87cm open])
  • Net weight: 126.8 lb. (57.5kg)
Refrigerated Model ST 16R
  • CFC-free refrigeration system
  • Temperature Range: -10° to +40°C
  • Pre-Cooling Function with direct button
  • 23.8L × 24.5W × 14.2 in.H [34.3 in.H open] (60.5 × 62.3 × 36cm [87cm open])
  • Net weight: 201.7 lb. (91.5kg)
Ordering Information:
A wide range of rotors are available for your daily routine processing requirements. Maximize performance with lighter-weight Thermo Scientific™ Fiberlite™ carbon fiber rotors that offer versatility, speed and robust, corrosion-free design. All rotors and adapters are sold separately.
Recommended for:
Cell Culture, Blood Processing
Specifications
Certifications/ComplianceIVD Compliant, IEC 61010-1, IEC 61010-2-202, IEC 61010-2-101. UL listed, CSA certified, CE marked, Certified Biosafety.
Controller TypeMicroprocessor
DescriptionSorvall ST 16 Centrifuge
Capacity4 x 400 mL
Max. RCF25,830 x g
Max. Speed15,200 rpm
Product LineSorvall
RefrigeratedNo
DisplayDigital
Drive SystemDirect, Brushless Induction Low Profile
Noise Level<61 dB(A)
Profile (Acceleration/Braking)9 Accel/10 Braking
Program Storage6 programs, direct access
RotorTX-400
Run Time9 hr. 99 min. plus HOLD
TypeVentilated Centrifuge
Electrical Requirements230 V 50/60 Hz